This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00928868_m1
See other SEMA3G GE Assays ›
Gene Symbol:
SEMA3G
Gene Name:
semaphorin 3G
Gene Aliases:
sem2
Chromosome Location:
Chr.3: 52433252 - 52445027 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00928868_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEMA3G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB029496.1 - 1 - 2 115 64
BC098104.1 - 1 - 2 115 40007797 64
BC098137.1 - 1 - 2 115 40007798 64
BC099724.1 - 1 - 2 115 40007801 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300011171.hg.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000231721.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000465657.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 ENST00000475739.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300011171.hg.1 NM_020163.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 108579 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0