Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022197.hg.1 |
ENST00000313050.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022197.hg.1 |
ENST00000313084.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022197.hg.1 |
ENST00000398348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022197.hg.1 |
ENST00000431893.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022197.hg.1 |
NM_014866.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022197.hg.1 |
TC09001732.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000277537.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000313050.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000313084.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000431893.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
NM_014866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000277537.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000313050.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000313084.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
ENST00000431893.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
NM_014866.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182080.hg.1 |
TC0900011980.hg.1 |
Clariom D TC |