This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00925641_m1
See other SNX29 GE Assays ›
Gene Symbol:
SNX29
Gene Name:
sorting nexin 29
Gene Aliases:
A-388D4.1, RUNDC2A
Chromosome Location:
Chr.16: 11976734 - 12574289 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00925641_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNX29
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032167.4 NP_115543.3 4 - 5 320 58
XM_005255682.3 XP_005255739.1 4 - 5 345 58
XM_005255683.3 XP_005255740.1 4 - 5 361 58
XM_011522738.2 XP_011521040.1 4 - 5 492 58
XM_011522741.2 XP_011521043.1 4 - 5 476 58
XM_011522743.2 XP_011521045.1 4 - 5 480 58
XM_011522744.2 XP_011521046.1 4 - 5 351 58
XM_017023871.1 XP_016879360.1 4 - 5 489 58
XM_017023872.1 XP_016879361.1 4 - 5 309 58
XM_017023873.1 XP_016879362.1 4 - 5 476 58
XM_017023874.1 XP_016879363.1 4 - 5 309 58
XM_017023876.1 XP_016879365.1 4 - 5 354 58
XM_017023877.1 XP_016879366.1 4 - 5 309 58
GenBank mRNA AK022425.1 - 4 - 5 309 58
AL135642.1 - 4 - 5 309 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600006941.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000167.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SNX29

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Process(es)

phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000562510.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000563308.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564111.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000564791.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000566228.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000568949.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 ENST00000569801.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600006941.hg.1 NM_032167.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0