Assay Details
Gene Symbol: ZNF567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300979.1 | NP_001287908.1 | 2 - 3 | 172 | 80 | |
NM_001322913.1 | NP_001309842.1 | 2 - 3 | 176 | 80 | ||
NM_001322916.1 | NP_001309845.1 | 2 - 3 | 120 | 80 | ||
NM_001322917.1 | NP_001309846.1 | 2 - 3 | 124 | 80 | ||
XM_017026417.1 | XP_016881906.1 | 4 - 5 | 1391 | 80 | ||
XM_017026418.1 | XP_016881907.1 | 2 - 3 | 251 | 80 | ||
GenBank mRNA | AK303326.1 | - | 2 - 3 | 156 | 80 | |
BX640706.1 | - | 2 - 3 | 161 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK303326.1,BX640706.1,NM_001300979.1,NM_001322913.1,NM_001322916.1,NM_001322917.1,XM_017026417.1,XM_017026418.1 transcripts.
This assay is in the 5'UTR region of the AK303326.1,BX640706.1,NM_001300979.1,NM_001322913.1,NM_001322916.1,NM_001322917.1,XM_017026417.1,XM_017026418.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
protein binding
transcription regulatory region DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159221.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159221.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159221.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159221.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159221.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159221.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159221.hg.1 | TC1900007950.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000360729.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000585696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000588311.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000589264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000590392.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | ENST00000591308.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007950.hg.1 | NM_152603.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159218.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159218.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159218.hg.1 | ENST00000536254.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900159218.hg.1 | NM_001300979.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900159218.hg.1 | TC1900007950.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 206994 | - | - |
