This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00925032_m1
See other PGBD5 GE Assays ›
Gene Symbol:
PGBD5
Gene Name:
piggyBac transposable element derived 5
Gene Aliases:
-
Chromosome Location:
Chr.1: 230321646 - 230425928 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00925032_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGBD5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258311.1 NP_001245240.1 5 - 6 1273 67
XM_017002330.1 XP_016857819.1 5 - 6 2342 67
GenBank mRNA AK297321.1 - 2 - 3 242 67
AK315968.1 - 2 - 3 303 67
AL833243.1 - 4 - 5 918 67
BC013901.2 - 1 - 2 82 67
BC027466.1 - 2 - 3 242 67
BC036865.1 - 4 - 5 700 67
BC150638.1 - 5 - 6 1090 9057008 67
KF670820.1 - 5 - 6 1074 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100017693.hg.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

viral or transposable element protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay PSR0100233503.hg.1 TC0100017693.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000391860.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000525115.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 ENST00000530424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq
Affymetrix Human Clariom D Assay TC0100017693.hg.1 NM_001258311.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0