Assay Details
Gene Symbol: SULT1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006588.3 | NP_006579.2 | 2 - 3 | 721 | 82 | |
XM_017003807.1 | XP_016859296.1 | 2 - 3 | 263 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000665.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017003807.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02010315.hg.1 | ENST00000272452.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02010315.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02010315.hg.1 | TC02000665.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02010317.hg.1 | ENST00000272452.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02010317.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02010317.hg.1 | TC02000665.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000409309.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008801.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200161387.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200161387.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200161387.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200161387.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200161387.hg.1 | TC0200008801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200161385.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200161385.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200161385.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200161385.hg.1 | ENST00000272452.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200161385.hg.1 | ENST00000494122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200161385.hg.1 | NM_006588.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200161385.hg.1 | TC0200008801.hg.1 | Clariom D TC |
