Assay Details
Gene Symbol: TMEM80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042463.2 | NP_001035928.2 | 3 - 4 | 373 | 62 | |
NM_001276274.1 | NP_001263203.1 | 3 - 4 | 373 | 62 | ||
NM_174940.3 | NP_777600.3 | 3 - 4 | 349 | 62 | ||
XM_006718206.3 | XP_006718269.3 | 3 - 4 | 379 | 62 | ||
XM_017017600.1 | XP_016873089.1 | 3 - 4 | 554 | 62 | ||
GenBank mRNA | AK302585.1 | - | 3 - 4 | 168 | 62 | |
BC008671.1 | - | 3 - 4 | 297 | 3854028 | 62 | |
BI916171.1 | - | 3 - 4 | 351 | 62 | ||
CF134831.1 | - | 2 - 3 | 139 | 62 | ||
CK819789.1 | - | 3 - 4 | 163 | 62 | ||
DR006597.1 | - | 3 - 4 | 316 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000025.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600009612.hg.1 | ENST00000619829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000528024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000528024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11000353.hg.1 | ENST00000397510.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11000353.hg.1 | ENST00000397512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11000353.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11000353.hg.1 | ENST00000526170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11000353.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11000353.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11000353.hg.1 | TC11000025.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100144796.hg.1 | TC1100006485.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600009612.hg.1 | ENST00000619829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397510.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000397512.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000488769.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000492023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000526170.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000528024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000528024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | ENST00000608174.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001042463.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276253.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_001276274.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006485.hg.1 | NM_174940.3 | Refseq |
