Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001155.hg.1 |
ENST00000215739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001155.hg.1 |
ENST00000389355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001155.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001155.hg.1 |
TC22001454.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001158.hg.1 |
ENST00000215739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001158.hg.1 |
ENST00000389355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001158.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001158.hg.1 |
TC22001454.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001156.hg.1 |
ENST00000215739.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001156.hg.1 |
ENST00000389355.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001156.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22001156.hg.1 |
TC22001454.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000215739.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000415354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000439171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000452988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000464807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000491432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000495142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000215739.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000415354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000439171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000452988.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000464807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000491432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
ENST00000495142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167397.hg.1 |
TC2200009196.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000215739.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000415354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000439171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000464807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000495142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000215739.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000415354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000439171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000464807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
ENST00000495142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167395.hg.1 |
TC2200009196.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000215739.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000415354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000439171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000464807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000491432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000495142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000215739.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000415354.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000439171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000464807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000491432.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
ENST00000495142.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
NM_006767.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200167394.hg.1 |
TC2200009196.hg.1 |
Clariom D TC |