This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00921603_m1
See other TRPM7 GE Assays ›
Gene Symbol:
TRPM7
Gene Name:
transient receptor potential cation channel subfamily M member 7
Gene Aliases:
ALSPDC, CHAK, CHAK1, LTRPC7, LTrpC-7, TRP-PLIK
Chromosome Location:
Chr.15: 50557158 - 50686835 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00921603_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRPM7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301212.1 NP_001288141.1 28 - 29 4760 200
XM_017022351.1 XP_016877840.1 29 - 30 4580 260
XM_017022353.1 XP_016877842.1 28 - 29 4520 200
GenBank mRNA AF346629.2 - 28 - 29 4716 200

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

cellular magnesium ion homeostasis
calcium-dependent cell-matrix adhesion
actomyosin structure organization
protein autophosphorylation
protein tetramerization
necroptotic process
calcium ion transmembrane transport
actin binding
protein serine/threonine kinase activity
calcium channel activity
ATP binding
myosin binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0