Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043726.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043726.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01043726.hg.1 |
TC01002807.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0100209479.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209479.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209479.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100209479.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100209479.hg.1 |
TC0100014685.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000370742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
ENST00000401034.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014685.hg.1 |
NM_022159.3 |
Refseq |