This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00920678_m1
See other ADGRL4 GE Assays ›
Gene Symbol:
ADGRL4
Gene Name:
adhesion G protein-coupled receptor L4
Gene Aliases:
ELTD1, ETL, KPG_003
Chromosome Location:
Chr.1: 78889764 - 79006810 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00920678_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022159.3 NP_071442.2 3 - 4 476 153
GenBank mRNA AB032017.1 - 3 - 4 437 153
AF192403.1 - 1 - 2 137 153
AK292894.1 - 3 - 4 369 153
AK313738.1 - 3 - 4 475 153
AY358360.1 - 3 - 4 343 153
BC025721.1 - 3 - 4 342 5229055 153

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014685.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002807.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRL4

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
calcium ion binding
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01043728.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01043728.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01043728.hg.1 TC01002807.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209481.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100209481.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209481.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100209481.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209481.hg.1 TC0100014685.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0