This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00920676_m1
See other ADGRL4 GE Assays ›
Gene Symbol:
ADGRL4
Gene Name:
adhesion G protein-coupled receptor L4
Gene Aliases:
ELTD1, ETL, KPG_003
Chromosome Location:
Chr.1: 78889764 - 79006810 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00920676_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022159.3 NP_071442.2 1 - 2 175 88
GenBank mRNA AB032017.1 - 1 - 2 136 88
AK292894.1 - 1 - 2 68 88
AK313738.1 - 1 - 2 174 88
AY358360.1 - 1 - 2 42 88
BC025721.1 - 1 - 2 41 5229055 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014685.hg
This assay targets the same location as HTA transcript cluster(s) TC01002807.hg

Additional Information:

This assay is in the 5'UTR region of the AB032017.1,AK292894.1,AK313738.1,AY358360.1,BC025721.1,NM_022159.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRL4

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
calcium ion binding
protein dimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01043730.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01043730.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01043730.hg.1 TC01002807.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01043731.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01043731.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01043731.hg.1 TC01002807.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01043733.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01043733.hg.1 TC01002807.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01043732.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01043732.hg.1 NM_022159.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01043732.hg.1 TC01002807.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100209486.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100209486.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209486.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100209486.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209486.hg.1 TC0100014685.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100209487.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209487.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209487.hg.1 TC0100014685.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 ENST00000401034.7 Ensembl
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay TC0100014685.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209483.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100209483.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209483.hg.1 ENST00000370742.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100209483.hg.1 NM_022159.3 Refseq
Affymetrix Human Clariom D Assay PSR0100209483.hg.1 TC0100014685.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 170254 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0