Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000177.hg.1 |
ENST00000381882.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000177.hg.1 |
ENST00000402452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000177.hg.1 |
ENST00000409241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000177.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000177.hg.1 |
TC20000015.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000176.hg.1 |
ENST00000381882.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000176.hg.1 |
ENST00000402452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000176.hg.1 |
ENST00000409241.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000176.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000176.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20000176.hg.1 |
TC20000015.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000169103.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169103.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169103.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169103.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169103.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169103.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169103.hg.1 |
TC2000009877.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000169104.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169104.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169104.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000169104.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000169104.hg.1 |
TC2000009877.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000246108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000409241.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
ENST00000477283.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009877.hg.1 |
NM_001136566.2 |
Refseq |