This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00918036_m1
See other OCRL GE Assays ›
Gene Symbol:
OCRL
Gene Name:
OCRL, inositol polyphosphate-5-phosphatase
Gene Aliases:
INPP5F, LOCR, NPHL2, OCRL-1, OCRL1
Chromosome Location:
Chr.X: 129532737 - 129592561 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00918036_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OCRL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000276.3 NP_000267.2 19 - 20 2305 106
NM_001318784.1 NP_001305713.1 19 - 20 2328 106
XM_005262422.2 XP_005262479.1 18 - 19 2112 106
XM_011531344.2 XP_011529646.1 20 - 21 2217 106
XM_011531345.2 XP_011529647.1 20 - 21 2220 106
XM_017029554.1 XP_016885043.1 19 - 20 2236 106
GenBank mRNA AK226116.1 - 19 - 20 2328 106
AK293107.1 - 19 - 20 2304 106
U57627.2 - 19 - 20 2305 106

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OCRL

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
lipid metabolic process
phosphatidylinositol biosynthetic process
signal transduction
cilium assembly
regulation of GTPase activity
positive regulation of GTPase activity
inositol phosphate metabolic process
phosphatidylinositol dephosphorylation
regulation of small GTPase mediated signal transduction
phosphatidylinositol-4,5-bisphosphate 5-phosphatase activity
GTPase activator activity
protein binding
Rac GTPase binding
inositol-1,4,5-trisphosphate 5-phosphatase activity
inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity
inositol phosphate phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0