This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00916455_m1
See other HS1BP3 GE Assays ›
Gene Symbol:
HS1BP3
Gene Name:
HCLS1 binding protein 3
Gene Aliases:
ETM2, HS1-BP3
Chromosome Location:
Chr.2: 20553861 - 20651661 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00916455_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HS1BP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022460.3 NP_071905.3 3 - 4 451 82
XM_017004696.1 XP_016860185.1 3 - 4 454 82
XM_017004697.1 XP_016860186.1 3 - 4 454 82
XM_017004698.1 XP_016860187.1 3 - 4 454 82
XM_017004699.1 XP_016860188.1 3 - 4 506 82
XM_017004700.1 XP_016860189.1 3 - 4 550 82
XM_017004701.1 XP_016860190.1 3 - 4 454 82
GenBank mRNA AK024311.1 - 3 - 4 444 82
AK314383.1 - 3 - 4 447 82
BC050636.1 - 3 - 4 431 5193161 82
BX354596.2 - 3 - 4 482 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200016640.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001625.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HS1BP3

Gene Ontology Categories:

Function(s) Process(es)

regulation of apoptotic process
phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02026015.hg.1 ENST00000304031.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026015.hg.1 ENST00000402541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026015.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02026015.hg.1 TC02001625.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000631166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 ENST00000631166.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay PSR0200225845.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200225845.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200225845.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay PSR0200225845.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200225845.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200225845.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay PSR0200225845.hg.1 TC0200016640.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000304031.7 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000402541.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000406618.3 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000445102.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000458740.5 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000631166.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 ENST00000631166.1 Ensembl
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq
Affymetrix Human Clariom D Assay TC0200016640.hg.1 NM_022460.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0