This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00914588_g1
See other ANXA7 GE Assays ›
Gene Symbol:
ANXA7
Gene Name:
annexin A7
Gene Aliases:
ANX7, SNX, SYNEXIN
Chromosome Location:
Chr.10: 73375101 - 73414085 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00914588_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ANXA7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001156.4 NP_001147.1 6 - 7 617 68
NM_001320879.1 NP_001307808.1 5 - 6 562 68
NM_001320880.1 NP_001307809.1 6 - 7 628 68
NM_004034.3 NP_004025.1 7 - 8 683 68
XM_017016162.1 XP_016871651.1 7 - 8 719 68
XM_017016163.1 XP_016871652.1 6 - 7 653 68
GenBank mRNA AB062429.1 - 6 - 7 575 68
AK222552.1 - 6 - 7 585 68
AK295319.1 - 6 - 7 494 68
AK300086.1 - 4 - 5 384 68
AK301681.1 - 6 - 7 371 68
AK307772.1 - 5 - 6 436 68
AK312447.1 - 7 - 8 656 68
AK313024.1 - 6 - 7 560 68
BC002632.2 - 6 - 7 577 3610541 68
BT007187.1 - 5 - 6 542 68
BX361177.2 - 5 - 6 528 68
CR407686.1 - 5 - 6 542 68
J04543.1 - 6 - 7 602 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011020.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001408.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ANXA7

Panther Classification:

Molecular Function -

calcium-binding protein

Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
autophagy
hemostasis
cell proliferation
regulation of cell shape
response to salt stress
cellular water homeostasis
negative regulation of gene expression
response to organic cyclic compound
epithelial cell differentiation
social behavior
response to calcium ion
membrane fusion
integrin binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
poly(A) RNA binding
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10017754.hg.1 ENST00000372919.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017754.hg.1 ENST00000372921.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017754.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017754.hg.1 ENST00000535178.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10017754.hg.1 NM_001156.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10017754.hg.1 NM_004034.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10017754.hg.1 TC10001408.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay PSR1000175036.hg.1 TC1000011020.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372919.8 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000372921.9 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000394847.3 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000463788.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 ENST00000492380.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_001156.4 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq
Affymetrix Human Clariom D Assay TC1000011020.hg.1 NM_004034.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0