This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00912257_m1
See other ARHGAP15 GE Assays ›
Gene Symbol:
ARHGAP15
Gene Name:
Rho GTPase activating protein 15
Gene Aliases:
BM046
Chromosome Location:
Chr.2: 143070973 - 143768352 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00912257_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018460.3 NP_060930.3 11 - 12 1156 80
XM_011511479.2 XP_011509781.1 12 - 13 1188 80
XM_011511481.2 XP_011509783.1 11 - 12 1091 80
XM_011511482.2 XP_011509784.1 11 - 12 1091 80
XM_011511483.1 XP_011509785.1 7 - 8 786 80
XM_017004499.1 XP_016859988.1 11 - 12 1117 80
XM_017004500.1 XP_016859989.1 10 - 11 983 80
GenBank mRNA AK226014.1 - 11 - 12 1091 80
BC016701.2 - 11 - 12 1062 4108644 80
BC038976.1 - 11 - 12 1061 5553184 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200009539.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000904.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARHGAP15

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of cell shape
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02013335.hg.1 ENST00000295095.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02013335.hg.1 NM_018460.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02013335.hg.1 TC02000904.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay PSR0200166672.hg.1 TC0200009539.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000295095.10 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000409869.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000419455.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000460776.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000469117.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000474474.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548242.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548800.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000548929.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549060.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000549436.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552289.1 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 ENST00000552641.5 Ensembl
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq
Affymetrix Human Clariom D Assay TC0200009539.hg.1 NM_018460.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0