Assay Details
Gene Symbol: ARHGAP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018460.3 | NP_060930.3 | 10 - 11 | 1079 | 72 | |
XM_011511479.2 | XP_011509781.1 | 11 - 12 | 1111 | 72 | ||
XM_011511481.2 | XP_011509783.1 | 10 - 11 | 1014 | 72 | ||
XM_011511482.2 | XP_011509784.1 | 10 - 11 | 1014 | 72 | ||
XM_011511483.1 | XP_011509785.1 | 6 - 7 | 709 | 72 | ||
XM_011511484.1 | XP_011509786.1 | 10 - 11 | 1014 | 72 | ||
XM_017004499.1 | XP_016859988.1 | 10 - 11 | 1040 | 72 | ||
XM_017004500.1 | XP_016859989.1 | 9 - 10 | 906 | 72 | ||
GenBank mRNA | AK226014.1 | - | 10 - 11 | 1014 | 72 | |
BC016701.2 | - | 10 - 11 | 985 | 4108644 | 72 | |
BC038976.1 | - | 10 - 11 | 984 | 5553184 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000904.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02013333.hg.1 | ENST00000295095.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02013333.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02013333.hg.1 | TC02000904.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000409869.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000460776.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000469117.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000474474.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548242.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548800.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000548929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549060.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552289.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | ENST00000552641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009539.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | ENST00000295095.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | ENST00000419455.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | ENST00000549436.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | NM_018460.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200166670.hg.1 | TC0200009539.hg.1 | Clariom D TC |
