This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00911935_g1
See other ADGRL1 GE Assays ›
Gene Symbol:
ADGRL1
Gene Name:
adhesion G protein-coupled receptor L1
Gene Aliases:
CIRL1, CL1, LEC2, LPHN1
Chromosome Location:
Chr.19: 14147737 - 14206245 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00911935_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008701.2 NP_001008701.1 22 - 23 3963 156
NM_014921.4 NP_055736.2 21 - 22 3948 156
XM_017026478.1 XP_016881967.1 22 - 23 4043 156
GenBank mRNA AB020628.1 - 22 - 23 3965 156
AF307079.1 - 21 - 22 3932 156
BC007587.1 - 12 - 13 1763 156

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRL1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
synaptic vesicle exocytosis
calcium-mediated signaling using intracellular calcium source
positive regulation of synapse assembly
positive regulation of synapse maturation
G-protein coupled receptor activity
protein binding
latrotoxin receptor activity
carbohydrate binding
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0