Assay Details
Gene Symbol: SLC28A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199633.1 | NP_001186562.1 | 14 - 15 | 1790 | 79 | |
NM_022127.2 | NP_071410.1 | 15 - 16 | 1736 | 79 | ||
NR_037638.2 | 15 - 16 | 1966 | 79 | |||
XM_011518905.2 | XP_011517207.1 | 14 - 15 | 1860 | 79 | ||
XM_011518906.2 | XP_011517208.1 | 15 - 16 | 1956 | 79 | ||
XM_011518907.2 | XP_011517209.1 | 13 - 14 | 1675 | 79 | ||
XM_011518908.2 | XP_011517210.1 | 9 - 10 | 1695 | 79 | ||
GenBank mRNA | AF305210.1 | - | 15 - 16 | 1736 | 79 | |
AK292849.1 | - | 14 - 15 | 1789 | 79 | ||
AK304406.1 | - | 15 - 16 | 1946 | 79 | ||
AK314039.1 | - | 14 - 15 | 1770 | 79 | ||
BC093821.1 | - | 14 - 15 | 1673 | 7939666 | 79 | |
BC093823.1 | - | 14 - 15 | 1673 | 7939668 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001286.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retina homeostasis
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
pyrimidine-containing compound transmembrane transport
purine nucleobase transmembrane transport
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity
retina homeostasis
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
pyrimidine-containing compound transmembrane transport
purine nucleobase transmembrane transport
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09016076.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09016076.hg.1 | ENST00000537648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09016076.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016076.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016076.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016076.hg.1 | TC09001286.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09016077.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09016077.hg.1 | ENST00000537648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09016077.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016077.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016077.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09016077.hg.1 | TC09001286.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172272.hg.1 | TC0900010591.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900172273.hg.1 | TC0900010591.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000376238.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | ENST00000495823.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_001199633.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NM_022127.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900010591.hg.1 | NR_037638.2 | Refseq |
