This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00910009_m1
See other GRID2 GE Assays ›
Gene Symbol:
GRID2
Gene Name:
glutamate ionotropic receptor delta type subunit 2
Gene Aliases:
GluD2, SCAR18
Chromosome Location:
Chr.4: 92304249 - 93810330 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00910009_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GRID2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286838.1 NP_001273767.1 13 - 14 2336 71
NM_001510.3 NP_001501.2 14 - 15 2621 71
XM_011531893.2 XP_011530195.2 14 - 15 8858 71
XM_011531894.2 XP_011530196.1 12 - 13 1897 71
XM_011531895.2 XP_011530197.1 12 - 13 1814 71
XM_017008118.1 XP_016863607.1 13 - 14 8810 71
XM_017008119.1 XP_016863608.1 14 - 15 2395 71
XM_017008120.1 XP_016863609.1 14 - 15 8858 71
XM_017008123.1 XP_016863612.1 12 - 13 3125 71
XM_017008124.1 XP_016863613.1 11 - 12 2345 71
XM_017008125.1 XP_016863614.1 11 - 12 1842 71
XM_017008126.1 XP_016863615.1 10 - 11 1558 71
GenBank mRNA AB209318.1 - 14 - 15 2621 71
AF009014.1 - 14 - 15 2363 71
BC099652.3 - 14 - 15 2363 40008396 71
BC099653.3 - 14 - 15 2363 40008397 71
BC099654.3 - 13 - 14 2078 40008398 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400008160.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000497.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GRID2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

transport
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
glutamate receptor signaling pathway
regulation of neuron projection development
cerebellar granule cell differentiation
ion transmembrane transport
cellular protein localization
ionotropic glutamate receptor signaling pathway
synaptic transmission, glutamatergic
regulation of neuron apoptotic process
excitatory postsynaptic potential
prepulse inhibition
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity
glutamate receptor activity
PDZ domain binding
scaffold protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000282020.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000502699.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000505687.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000510992.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000512631.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000513976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000515744.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 ENST00000611049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001286838.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008160.hg.1 NM_001510.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0