Assay Details
Gene Symbol: ZC3H7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017590.5 | NP_060060.3 | 7 - 8 | 858 | 100 | |
XM_011530016.2 | XP_011528318.1 | 7 - 8 | 839 | 100 | ||
GenBank mRNA | AB028954.1 | - | 4 - 5 | 471 | 100 | |
AK225891.1 | - | 7 - 8 | 831 | 100 | ||
AK315009.1 | - | 7 - 8 | 800 | 100 | ||
BC152558.1 | - | 7 - 8 | 836 | 8860405 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000335.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011530016.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006441.hg.1 | ENST00000351589.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006441.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006441.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006441.hg.1 | TC22000335.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200152783.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152783.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152783.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152783.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152783.hg.1 | TC2200007467.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000352645.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | ENST00000486331.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007467.hg.1 | NM_017590.5 | Refseq |
