This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00909349_m1
See other SIPA1L2 GE Assays ›
Gene Symbol:
SIPA1L2
Gene Name:
signal induced proliferation associated 1 like 2
Gene Aliases:
SPAL2
Chromosome Location:
Chr.1: 232397965 - 232630491 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00909349_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIPA1L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020808.4 NP_065859.3 6 - 7 2505 98
XM_005273211.2 XP_005273268.1 8 - 9 2842 98
XM_005273212.4 XP_005273269.1 8 - 9 2649 98
XM_005273213.4 XP_005273270.1 7 - 8 2542 98
XM_011544243.2 XP_011542545.1 9 - 10 2698 98
XM_017001896.1 XP_016857385.1 8 - 9 2595 98
XM_017001897.1 XP_016857386.1 8 - 9 2842 98
XM_017001898.1 XP_016857387.1 8 - 9 2616 98
GenBank mRNA AB037810.1 - 6 - 7 1709 98
AY168879.1 - 6 - 7 2465 98
BC150334.1 - 6 - 7 1720 8860510 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SIPA1L2

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000486472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000486472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000486472.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000494056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000494056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000494056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000495863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000495863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 ENST00000495863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000262861.8 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000308942.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000366630.5 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000486472.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000486472.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000486472.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000494056.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000494056.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000494056.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000495863.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000495863.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 ENST00000495863.1 Ensembl
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq
Affymetrix Human Clariom D Assay TC0100017748.hg.1 NM_020808.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0