This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00909271_m1
See other IL1RL2 GE Assays ›
Gene Symbol:
IL1RL2
Gene Name:
interleukin 1 receptor like 2
Gene Aliases:
IL-1Rrp2, IL-36R, IL1R-rp2, IL1RRP2
Chromosome Location:
Chr.2: 102186973 - 102242906 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00909271_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL1RL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003854.2 NP_003845.2 1 - 2 115 57
XM_005264035.1 XP_005264092.1 1 - 2 108 57
XM_011512091.1 XP_011510393.1 1 - 2 108 57
XM_011512092.1 XP_011510394.1 1 - 2 108 57
XM_011512093.1 XP_011510395.1 1 - 2 108 57
XM_011512096.1 XP_011510398.1 1 - 2 108 57
XM_017005174.1 XP_016860663.1 1 - 2 108 57
XM_017005178.1 XP_016860667.1 1 - 2 57 57
XM_017005179.1 XP_016860668.1 1 - 2 103 57
GenBank mRNA AF284434.1 - 1 - 2 123 57
U49065.1 - 1 - 2 123 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200008669.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000621.hg.1

Additional Information:

This assay is in the 5'UTR region of the AF284434.1,NM_003854.2,U49065.1,XM_017005178.1,XM_017005179.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL1RL2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
cellular defense response
signal transduction
positive regulation of interleukin-6 production
innate immune response
positive regulation of T cell differentiation
regulation of inflammatory response
interleukin-1-mediated signaling pathway
interleukin-1 receptor activity
interleukin-1, Type I, activating receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02009858.hg.1 ENST00000264257.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009858.hg.1 ENST00000441515.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009858.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009858.hg.1 ENST00000539491.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009858.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02009858.hg.1 TC02000621.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02009855.hg.1 ENST00000264257.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009855.hg.1 ENST00000441515.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009855.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009855.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02009855.hg.1 TC02000621.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02009854.hg.1 ENST00000264257.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02009854.hg.1 NM_003854.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02009854.hg.1 TC02000621.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160537.hg.1 TC0200008669.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay TC0200008669.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160535.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160535.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160535.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160535.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160535.hg.1 TC0200008669.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000264257.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000421464.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000441515.3 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 ENST00000481806.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 NM_003854.3 Refseq
Affymetrix Human Clariom D Assay PSR0200160541.hg.1 TC0200008669.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0