This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00909201_g1
See other ADGRL1 GE Assays ›
Gene Symbol:
ADGRL1
Gene Name:
adhesion G protein-coupled receptor L1
Gene Aliases:
CIRL1, CL1, LEC2, LPHN1
Chromosome Location:
Chr.19: 14147737 - 14206245 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00909201_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008701.2 NP_001008701.1 17 - 18 3345 89
NM_014921.4 NP_055736.2 16 - 17 3330 89
XM_005259818.3 XP_005259875.1 16 - 17 3386 89
XM_011527796.2 XP_011526098.1 17 - 18 3422 89
XM_011527798.2 XP_011526100.1 16 - 17 3407 89
XM_011527801.2 XP_011526103.1 15 - 16 2862 89
XM_017026475.1 XP_016881964.1 17 - 18 3425 89
XM_017026476.1 XP_016881965.1 18 - 19 3535 89
XM_017026477.1 XP_016881966.1 17 - 18 3404 89
XM_017026478.1 XP_016881967.1 17 - 18 3425 89
XM_017026479.1 XP_016881968.1 17 - 18 3425 89
XM_017026480.1 XP_016881969.1 14 - 15 2840 89
GenBank mRNA AB020628.1 - 17 - 18 3347 89
AF307079.1 - 16 - 17 3314 89
BC007587.1 - 7 - 8 1145 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900009808.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001233.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRL1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
synaptic vesicle exocytosis
calcium-mediated signaling using intracellular calcium source
positive regulation of synapse assembly
positive regulation of synapse maturation
G-protein coupled receptor activity
protein binding
latrotoxin receptor activity
carbohydrate binding
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19017339.hg.1 ENST00000340736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19017339.hg.1 ENST00000361434.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19017339.hg.1 NM_001008701.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19017339.hg.1 NM_014921.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19017339.hg.1 TC19001233.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 ENST00000340736.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 ENST00000361434.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 NM_001008701.2 Refseq
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 NM_014921.4 Refseq
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 ENST00000340736.10 Ensembl
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 ENST00000361434.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 NM_001008701.2 Refseq
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 NM_014921.4 Refseq
Affymetrix Human Clariom D Assay PSR1900180287.hg.1 TC1900009808.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0