Assay Details
Gene Symbol: DHTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018706.6 | NP_061176.3 | 3 - 4 | 640 | 73 | |
GenBank mRNA | AK309810.1 | - | 3 - 4 | 548 | 73 | |
BC002477.2 | - | 3 - 4 | 585 | 3347913 | 73 | |
BC007955.2 | - | 3 - 4 | 567 | 4302613 | 73 | |
CF593518.1 | - | 3 - 4 | 652 | 73 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000089.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10000923.hg.1 | ENST00000263035.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10000923.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10000923.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10000923.hg.1 | TC10000089.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000146360.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000146360.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000146360.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000146360.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000146360.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000146360.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000146360.hg.1 | TC1000006794.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000263035.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000415935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000437298.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000448829.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000465617.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | ENST00000479283.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006794.hg.1 | NM_018706.6 | Refseq |
