Assay Details
Gene Symbol: ERC2-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024615.1 | 1 | 2203 | 86 | ||
GenBank mRNA | U88965.1 | - | 1 | 2114 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ELKS/RAB6-interacting/CAST family member 2 | ERC2 |
CAST, CAST1, ELKSL, SPBC110, Spc110 |
Sense | Chr. 3 - 55508311 - 56468473 on NCBI Build GRCh38 | |
Downstream | microRNA 3938 | MIR3938 | 193.1kb | Sense | Chr. 3 - 55852492 - 55852594 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P18055 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P18053 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P18055 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 43543 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 38100 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4927 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P18055 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3537861 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P18055 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256407 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256376 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256408 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256369 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256386 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391458 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256380 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391421 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391463 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15441515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256379 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256367 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256374 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256365 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256412 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256414 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256364 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256374 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391458 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256400 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256401 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391421 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256408 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391442 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391429 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256386 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391463 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256406 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391437 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256407 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256380 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256369 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256376 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256371 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256412 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15441515 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256426 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391424 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22391432 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256367 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256379 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256414 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256365 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256410 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02256364 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P18053 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P18055 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P18055 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P18053 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256371 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256420 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256415 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256369 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256390 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15441515 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15441514 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256412 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256406 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256397 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256404 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256378 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15441512 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256382 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256376 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256408 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256428 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15441517 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256387 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256401 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256374 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256417 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256407 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256434 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256423 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256385 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256367 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256410 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256394 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P02256364 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8088216 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8088217 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8088215 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8088213 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8088218 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8088214 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8088212 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208247_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11750318_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11750318_at | - | - |
Human Genome U133 Plus 2.0 Array | 208247_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208247_at | - | - |
Human Genome U133A 2.0 Array | 208247_at | - | - |
Human Genome U133A 2.0 Array | 208247_PM_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw553922 | - | - |
Human Genome U95 Set | 31392_PM_r_at | - | - |
Human Genome U95 Set | 31392_r_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208247_at3 | - | - |
Human Genome U133 Set | 208247_at | - | - |
Human Genome U133 Set | 208247_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021177 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021177 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 137390 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3242335 | U88965.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3242335 | NR_024615.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3242335 | U88965.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3242335 | NR_024615.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242335 | U88965.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242335 | NR_024615.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3242335 | U88965.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3242335 | NR_024615.1 | human_transcriptome |
