This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00907248_m1
See other S100A5 GE Assays ›
Gene Symbol:
S100A5
Gene Name:
S100 calcium binding protein A5
Gene Aliases:
S100D
Chromosome Location:
Chr.1: 153537114 - 153543514 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00907248_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002962.1 NP_002953.2 2 - 3 272 80
XM_017002029.1 XP_016857518.1 2 - 3 366 80
XM_017002030.1 XP_016857519.1 2 - 3 695 80
GenBank mRNA BC093955.1 - 2 - 3 128 7939800 80
BC093957.2 - 2 - 3 128 7939802 80
Z18954.1 - 2 - 3 272 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100015865.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003265.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_002962.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100A5

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

copper ion binding
calcium ion binding
zinc ion binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01050243.hg.1 ENST00000359215.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01050243.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01050243.hg.1 ENST00000368718.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01050243.hg.1 NM_002962.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01050243.hg.1 TC01003265.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100219156.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100219156.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100219156.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay PSR0100219156.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100219156.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100219156.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay PSR0100219156.hg.1 TC0100015865.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0