This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00907150_m1
See other GUCY1A2 GE Assays ›
Gene Symbol:
GUCY1A2
Gene Name:
guanylate cyclase 1 soluble subunit alpha 2
Gene Aliases:
GC-SA2, GUC1A2
Chromosome Location:
Chr.11: 106674012 - 107018445 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00907150_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GUCY1A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000855.2 NP_000846.1 3 - 4 879 120
NM_001256424.1 NP_001243353.1 3 - 4 879 120
GenBank mRNA BC130484.1 - 3 - 4 585 40146515 120
BC130488.1 - 3 - 4 585 40146519 120
BC144033.1 - 3 - 4 585 9052553 120
X63282.1 - 3 - 4 879 120
Z50053.1 - 3 - 4 879 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012197.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11002258.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GUCY1A2

Panther Classification:

Molecular Function -

guanylate cyclase

Gene Ontology Categories:

Function(s) Process(es)

cGMP biosynthetic process
signal transduction
positive regulation of cGMP biosynthetic process
intracellular signal transduction
adenylate cyclase activity
guanylate cyclase activity
GTP binding
heme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11026997.hg.1 ENST00000282249.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11026997.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11026997.hg.1 ENST00000526355.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11026997.hg.1 NM_000855.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11026997.hg.1 NM_001256424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11026997.hg.1 TC11002258.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay PSR1100193810.hg.1 TC1100012197.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000282249.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000347596.2 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 ENST00000526355.6 Ensembl
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_000855.2 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq
Affymetrix Human Clariom D Assay TC1100012197.hg.1 NM_001256424.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0