Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018972.hg.1 |
ENST00000251636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018972.hg.1 |
NM_019030.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018972.hg.1 |
TC05001362.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018973.hg.1 |
ENST00000251636.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018973.hg.1 |
NM_019030.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018973.hg.1 |
TC05001362.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
ENST00000251636.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
ENST00000504778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
ENST00000621106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
NM_019030.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
ENST00000251636.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
ENST00000504778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
ENST00000621106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
NM_019030.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175066.hg.1 |
TC0500010760.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
ENST00000251636.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
ENST00000504778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
ENST00000621106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
NM_019030.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
ENST00000251636.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
ENST00000504778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
ENST00000621106.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
NM_019030.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500175068.hg.1 |
TC0500010760.hg.1 |
Clariom D TC |