Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
ENST00000313698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
ENST00000392636.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
ENST00000413681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
ENST00000455480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
ENST00000508303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
NM_001136538.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
NM_025247.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011437.hg.1 |
TC12003211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
ENST00000313698.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
ENST00000392636.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
ENST00000413681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
ENST00000455480.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
ENST00000508303.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
NM_001136538.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
NM_025247.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12011435.hg.1 |
TC12003211.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000313698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000413681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000455480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000504803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000505487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000507688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000508303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000550198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
NM_001136538.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
NM_025247.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000313698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000413681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000455480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000504803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000505487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000507688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000508303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
ENST00000550198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
NM_001136538.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
NM_025247.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201610.hg.1 |
TC1200012706.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000313698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000413681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000455480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000504803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000505487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000508303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000546647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000550198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
NM_001136538.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
NM_025247.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000313698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000413681.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000455480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000504803.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000505487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000508303.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000546647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
ENST00000550198.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
NM_001136538.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
NM_025247.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200201607.hg.1 |
TC1200012706.hg.1 |
Clariom D TC |