This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00905343_m1
See other SVOP GE Assays ›
Gene Symbol:
SVOP
Gene Name:
SV2 related protein
Gene Aliases:
-
Chromosome Location:
Chr.12: 108908129 - 109021240 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00905343_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SVOP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018711.4 NP_061181.1 9 - 10 1267 107
GenBank mRNA BC033587.1 - 9 - 10 1067 5243017 107
BC094722.1 - 9 - 10 914 4817829 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200011855.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001943.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SVOP

Gene Ontology Categories:

Function(s) Process(es)

transmembrane transport
substrate-specific transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000551683.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000551683.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000551683.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12025293.hg.1 ENST00000299134.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025293.hg.1 NM_018711.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12025293.hg.1 TC12001943.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200191940.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200191940.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200191940.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay PSR1200191940.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200191940.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200191940.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay PSR1200191940.hg.1 TC1200011855.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000546618.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000548229.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000550436.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000551211.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000551683.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000551683.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000551683.2 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000569754.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 ENST00000610966.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq
Affymetrix Human Clariom D Assay TC1200011855.hg.1 NM_018711.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0