This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00904068_m1
See other PIK3C2A GE Assays ›
Gene Symbol:
PIK3C2A
Gene Name:
phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha
Gene Aliases:
CPK, PI3-K-C2(ALPHA), PI3-K-C2A, PI3K-C2-alpha, PI3K-C2alpha
Chromosome Location:
Chr.11: 17086572 - 17207996 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00904068_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIK3C2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321378.1 NP_001308307.1 10 - 11 2133 109
NM_001321380.1 NP_001308309.1 8 - 9 934 109
NM_002645.3 NP_002636.2 9 - 10 2064 109
XM_011520186.2 XP_011518488.1 9 - 10 2059 109
XM_017017924.1 XP_016873413.1 10 - 11 2080 109
GenBank mRNA AK294416.1 - 8 - 9 921 109
AK299474.1 - 1996 109
BC113658.1 - 8 - 9 1916 8322710 109
Y13367.1 - 8 - 9 1850 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013148.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003476.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PIK3C2A

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylinositol biosynthetic process
exocytosis
endocytosis
epidermal growth factor receptor signaling pathway
insulin receptor signaling pathway
vascular smooth muscle contraction
phosphatidylinositol-3-phosphate biosynthetic process
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
clathrin coat assembly
ATP binding
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol 3-kinase activity
1-phosphatidylinositol-4-phosphate 3-kinase activity
phosphatidylinositol binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000503729.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000503729.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000503729.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000532035.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000532035.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000532035.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018281.hg.1 ENST00000265970.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018281.hg.1 ENST00000540361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018281.hg.1 NM_002645.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018281.hg.1 TC11003476.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100203731.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100203731.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay PSR1100203731.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100203731.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay PSR1100203731.hg.1 TC1100013148.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000265970.11 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000503729.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000503729.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000503729.2 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000532035.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000532035.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000532035.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 ENST00000533645.1 Ensembl
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq
Affymetrix Human Clariom D Assay TC1100013148.hg.1 NM_002645.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0