This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00903546_m1
See other SDR42E1 GE Assays ›
Gene Symbol:
SDR42E1
Gene Name:
short chain dehydrogenase/reductase family 42E, member 1
Gene Aliases:
HSPC105
Chromosome Location:
Chr.16: 81988855 - 82011488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00903546_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDR42E1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005256257.3 XP_005256314.1 2 - 3 310 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005256257.3 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
oxidation-reduction process
3-beta-hydroxy-delta5-steroid dehydrogenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000328945.6 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000532128.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 ENST00000534209.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq
Affymetrix Human Clariom D Assay TC1600011002.hg.1 NM_145168.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0