This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00902979_m1
See other S100B GE Assays ›
Gene Symbol:
S100B
Gene Name:
S100 calcium binding protein B
Gene Aliases:
NEF, S100, S100-B, S100beta
Chromosome Location:
Chr.21: 46598618 - 46605242 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00902979_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017028424.1 XP_016883913.1 1 - 2 233 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017028424.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

axonogenesis
central nervous system development
learning or memory
memory
cell proliferation
positive regulation of cell proliferation
regulation of cell shape
positive regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
regulation of neuronal synaptic plasticity
astrocyte differentiation
response to glucocorticoid
response to methylmercury
long-term synaptic potentiation
cellular response to hypoxia
negative regulation of skeletal muscle cell differentiation
calcium ion binding
protein binding
zinc ion binding
identical protein binding
protein homodimerization activity
S100 protein binding
tau protein binding
calcium-dependent protein binding
RAGE receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000291700.8 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000367071.4 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 ENST00000397648.1 Ensembl
Affymetrix Human Clariom D Assay TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Clariom D Assay TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Clariom D Assay TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Clariom D Assay TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Clariom D Assay TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Clariom D Assay TC2100008477.hg.1 NM_006272.2 Refseq
Affymetrix Human Clariom D Assay TC2100008477.hg.1 NM_006272.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0