This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00902906_m1
See other MSANTD3-TMEFF1 GE Assays ›
Gene Symbol:
MSANTD3-TMEFF1 TMEFF1
Gene Name:
MSANTD3-TMEFF1 readthrough
transmembrane protein with EGF like and two follistatin like domains 1
Gene Aliases:
C9orf3-TMEFF1, C9orf30-TMEFF1
C9orf2, CT120.1, H7365, TR-1
Chromosome Location:
Chr.9: 100441906 - 100577636 on Build GRCh38
Chromosome Location:
Chr.9: 100473238 - 100577636 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs00902906_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD3-TMEFF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198812.1 NP_001185741.1 2 - 3 563 136
GenBank mRNA AL831919.1 - 2 - 3 198 136
Gene Symbol: TMEFF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003692.4 NP_003683.2 2 - 3 615 136
GenBank mRNA BC035056.1 - 2 - 3 417 5262325 136

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900012154.hg.1, TC0900012155.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09002903.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MSANTD3-TMEFF1
Click here to find siRNAs for Gene TMEFF1

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s)

multicellular organism development

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09005627.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09005627.hg.1 NM_003692.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09005627.hg.1 TC09002903.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900184349.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900184349.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay PSR0900184349.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay PSR0900184349.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay PSR0900184349.hg.1 TC0900012154.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900184363.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184363.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184363.hg.1 TC0900012155.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay TC0900012155.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184364.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184364.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184364.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184364.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184364.hg.1 TC0900012155.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900184366.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184366.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184366.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184366.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184366.hg.1 TC0900012155.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900184365.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184365.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184365.hg.1 ENST00000502978.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900184365.hg.1 NM_001198812.1 Refseq
Affymetrix Human Clariom D Assay PSR0900184365.hg.1 TC0900012155.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 ENST00000374879.4 Ensembl
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq
Affymetrix Human Clariom D Assay TC0900012154.hg.1 NM_003692.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0