This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00902146_m1
See other TUBGCP3 GE Assays ›
Gene Symbol:
TUBGCP3
Gene Name:
tubulin gamma complex associated protein 3
Gene Aliases:
104p, GCP3, Grip104, SPBC98, Spc98, Spc98p
Chromosome Location:
Chr.13: 112485005 - 112606469 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00902146_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TUBGCP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001286277.1 NP_001273206.1 8 - 9 1143 72
NM_001286278.1 NP_001273207.1 8 - 9 1173 72
NM_001286279.1 NP_001273208.1 8 - 9 1173 72
NM_006322.5 NP_006313.1 8 - 9 1173 72
XM_005268293.1 XP_005268350.1 8 - 9 1168 72
XM_011537462.2 XP_011535764.1 8 - 9 1126 72
XM_011537463.2 XP_011535765.1 9 - 10 1102 72
XM_011537464.2 XP_011535766.1 8 - 9 1906 72
XM_017020323.1 XP_016875812.1 9 - 10 1520 72
GenBank mRNA AF042378.1 - 8 - 9 1052 72
AJ003061.1 - 8 - 9 968 72
AJ003062.1 - 8 - 9 968 72
AK302537.1 - 8 - 9 1090 72
AK314785.1 - 8 - 9 1173 72
BC007763.2 - 8 - 9 1123 72
BC013781.1 - 8 - 9 1113 3867019 72
BC046634.2 - 8 - 9 1156 5295831 72
BT009772.1 - 8 - 9 968 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009856.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000887.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TUBGCP3

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule nucleation
meiotic nuclear division
single fertilization
cytoplasmic microtubule organization
centrosome duplication
interphase microtubule nucleation by interphase microtubule organizing center
mitotic spindle assembly
structural molecule activity
structural constituent of cytoskeleton
protein binding
gamma-tubulin binding
microtubule minus-end binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000261965.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000375669.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000462580.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000464139.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000469302.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000483532.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000611152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 ENST00000618439.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286278.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_001286279.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009856.hg.1 NM_006322.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0