This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00901597_g1
See other S100A5 GE Assays ›
Gene Symbol:
S100A5
Gene Name:
S100 calcium binding protein A5
Gene Aliases:
S100D
Chromosome Location:
Chr.1: 153537114 - 153543514 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00901597_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100A5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011509860.2 XP_011508162.1 1 - 2 759 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011509860.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Process(es)

copper ion binding
calcium ion binding
zinc ion binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368717.2 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 ENST00000368718.5 Ensembl
Affymetrix Human Clariom D Assay TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay TC0100015865.hg.1 NM_002962.1 Refseq
Affymetrix Human Clariom D Assay TC0100015865.hg.1 NM_002962.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0