This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00898192_g1
See other TM9SF4 GE Assays ›
Gene Symbol:
TM9SF4
Gene Name:
transmembrane 9 superfamily member 4
Gene Aliases:
dJ836N17.2
Chromosome Location:
Chr.20: 32109506 - 32167258 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00898192_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TM9SF4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014742.3 NP_055557.2 14 - 15 1742 93
XM_005260622.4 XP_005260679.1 14 - 15 1794 93
XM_017028154.1 XP_016883643.1 13 - 14 1439 93
XM_017028155.1 XP_016883644.1 13 - 14 1710 93
XM_017028156.1 XP_016883645.1 14 - 15 1490 93
XM_017028157.1 XP_016883646.1 14 - 15 1747 93
XM_017028158.1 XP_016883647.1 13 - 14 1401 93
XM_017028159.1 XP_016883648.1 14 - 15 1547 93
XM_017028160.1 XP_016883649.1 14 - 15 1459 93
XM_017028161.1 XP_016883650.1 13 - 14 1411 93
GenBank mRNA AK126534.1 - 12 - 13 1192 93
AK294980.1 - 14 - 15 1490 93
AK296500.1 - 14 - 15 1399 93
AK316293.1 - 13 - 14 1446 93
BC021107.1 - 14 - 15 1518 4896167 93
BC022850.2 - 13 - 14 1500 93
D87444.1 - 14 - 15 1796 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007105.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000207.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TM9SF4

Panther Classification:

Molecular Function -

transporter

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20002549.hg.1 ENST00000217315.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20002549.hg.1 ENST00000398022.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20002549.hg.1 NM_014742.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20002549.hg.1 TC20000207.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2000148564.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay PSR2000148564.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000148564.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay PSR2000148564.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay PSR2000148564.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay PSR2000148564.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay PSR2000148564.hg.1 TC2000007105.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000217315.9 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000398022.6 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000417389.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000442842.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000450829.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000479591.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 ENST00000495749.1 Ensembl
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq
Affymetrix Human Clariom D Assay TC2000007105.hg.1 NM_014742.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header