This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00893064_m1
See other PPFIA3 GE Assays ›
Gene Symbol:
PPFIA3
Gene Name:
PTPRF interacting protein alpha 3
Gene Aliases:
LPNA3
Chromosome Location:
Chr.19: 49119389 - 49151030 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00893064_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPFIA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003660.3 NP_003651.1 23 - 24 3185 57
NR_103842.1 22 - 23 2962 57
GenBank mRNA AB014554.2 - 23 - 24 3168 57
AF034800.1 - 6 - 7 583 57
AK289757.1 - 23 - 24 2890 57
BC101518.1 - 23 - 24 2911 8069024 57
BC101520.1 - 23 - 24 2911 8069026 57
BC143470.1 - 22 - 23 2688 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900008527.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000727.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPFIA3

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter secretion
glutamate secretion
regulation of short-term neuronal synaptic plasticity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19010080.hg.1 ENST00000334186.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19010080.hg.1 NM_003660.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19010080.hg.1 TC19000727.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000334186.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602351.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602655.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602848.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602897.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 NM_003660.3 Refseq
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 NR_103842.1 Refseq
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000334186.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602351.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602655.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602848.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 ENST00000602897.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 NM_003660.3 Refseq
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 NR_103842.1 Refseq
Affymetrix Human Clariom D Assay PSR1900165728.hg.1 TC1900008527.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0