Assay Details
Gene Symbol: FCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK309540.1 | - | 3 - 4 | 234 | 64 | |
BG546674.1 | - | 3 - 4 | 267 | 64 | ||
CR456808.1 | - | 3 - 4 | 232 | 64 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000270879.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000354982.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000481748.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | ENST00000498393.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_003665.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100013429.hg.1 | NM_173452.2 | Refseq |
