Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011207.hg.1 |
ENST00000457453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166373.hg.1 |
TC0500009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011207.hg.1 |
ENST00000457453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166370.hg.1 |
TC0500009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500166371.hg.1 |
TC0500009481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000265093.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000517669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519374.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
ENST00000519911.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009481.hg.1 |
NM_003945.3 |
Refseq |