This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00827104_m1
See other CCBE1 GE Assays ›
Gene Symbol:
CCBE1
Gene Name:
collagen and calcium binding EGF domains 1
Gene Aliases:
HKLLS1
Chromosome Location:
Chr.18: 59430939 - 59698181 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00827104_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCBE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133459.3 NP_597716.1 7 - 8 846 86
XM_017025558.1 XP_016881047.1 7 - 8 1614 86
GenBank mRNA AB075863.1 - 7 - 8 814 86
AY358347.1 - 7 - 8 792 86
BC046645.1 - 2 - 3 481 5766933 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800008827.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000548.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CCBE1

Panther Classification:

Molecular Function -

extracellular matrix structural protein

Gene Ontology Categories:

Function(s) Process(es)

lymphangiogenesis
sprouting angiogenesis
positive regulation of vascular endothelial growth factor production
positive regulation of endothelial cell migration
positive regulation of protein processing
positive regulation of angiogenesis
venous blood vessel morphogenesis
positive regulation of vascular endothelial growth factor signaling pathway
positive regulation of lymphangiogenesis
protease binding
calcium ion binding
protein binding
collagen binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005693.hg.1 ENST00000439986.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005693.hg.1 NM_133459.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005693.hg.1 TC18000548.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 ENST00000589419.1 Ensembl
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay TC1800008827.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay PSR1800160786.hg.1 TC1800008827.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 ENST00000398179.3 Ensembl
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 ENST00000439986.8 Ensembl
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 ENST00000589116.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 NM_133459.3 Refseq
Affymetrix Human Clariom D Assay PSR1800160787.hg.1 TC1800008827.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0