This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00765773_m1
See other ANGPTL2 GE Assays ›
Gene Symbol:
ANGPTL2
Gene Name:
angiopoietin like 2
Gene Aliases:
ARP2, HARP
Chromosome Location:
Chr.9: 127087349 - 127122803 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00765773_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ANGPTL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012098.2 NP_036230.1 1 - 2 456 74
XM_006717030.3 XP_006717093.1 1 - 2 494 74
GenBank mRNA AK307175.1 - 1 - 2 352 74
AK312876.1 - 1 - 2 375 74
AL598400.1 - 1 - 2 457 74
AY358274.1 - 1 - 2 475 74
BC012368.1 - 1 - 2 329 3865062 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011565.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001599.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK312876.1,AY358274.1,BC012368.1,NM_012098.2,XM_006717030.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ANGPTL2

Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
receptor binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09020066.hg.1 ENST00000373425.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09020066.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09020066.hg.1 TC09001599.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000470194.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000470194.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000491991.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 ENST00000491991.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09020067.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09020067.hg.1 ENST00000373425.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09020067.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09020067.hg.1 TC09001599.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09020069.hg.1 ENST00000373425.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09020069.hg.1 NM_012098.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09020069.hg.1 TC09001599.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900178808.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178808.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178808.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178808.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178808.hg.1 TC0900011565.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900178809.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178809.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178809.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178809.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178809.hg.1 TC0900011565.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000470194.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000470194.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000491991.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 ENST00000491991.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay TC0900011565.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178812.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178812.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178812.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178812.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178812.hg.1 TC0900011565.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900178810.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900178810.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178810.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178810.hg.1 ENST00000373417.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900178810.hg.1 ENST00000373425.7 Ensembl
Affymetrix Human Clariom D Assay PSR0900178810.hg.1 NM_012098.2 Refseq
Affymetrix Human Clariom D Assay PSR0900178810.hg.1 TC0900011565.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 189705 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0