Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036354.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036354.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036354.hg.1 |
TC02002256.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202849.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202849.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202849.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202849.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202849.hg.1 |
TC0200014049.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014049.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202848.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202848.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202848.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202848.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202848.hg.1 |
TC0200014049.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200202850.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202850.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202850.hg.1 |
ENST00000426077.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202850.hg.1 |
NM_024121.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202850.hg.1 |
TC0200014049.hg.1 |
Clariom D TC |