This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00699604_m1
See other SLC6A18 GE Assays ›
Gene Symbol:
SLC6A18
Gene Name:
solute carrier family 6 member 18
Gene Aliases:
Xtrp2
Chromosome Location:
Chr.5: 1225355 - 1246189 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00699604_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC6A18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182632.2 NP_872438.2 3 - 4 561 124
GenBank mRNA AK055798.1 - 3 - 4 561 124
BC056757.1 - 3 - 4 519 5758770 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500006494.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000017.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC6A18

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
neurotransmitter transport
amino acid transport
neurotransmitter:sodium symporter activity
amino acid transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05000342.hg.1 ENST00000296821.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05000342.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05000342.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05000342.hg.1 TC05000017.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05000343.hg.1 ENST00000296821.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05000343.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05000343.hg.1 NM_182632.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05000343.hg.1 TC05000017.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 ENST00000513607.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay TC0500006494.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay PSR0500144685.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500144685.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay PSR0500144685.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500144685.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay PSR0500144685.hg.1 TC0500006494.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500144686.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500144686.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay PSR0500144686.hg.1 ENST00000324642.3 Ensembl
Affymetrix Human Clariom D Assay PSR0500144686.hg.1 NM_182632.2 Refseq
Affymetrix Human Clariom D Assay PSR0500144686.hg.1 TC0500006494.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0