Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000465815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000465815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001707.hg.1 |
ENST00000255006.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001707.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001707.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001707.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001707.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20001707.hg.1 |
TC20000134.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000146950.hg.1 |
TC2000006861.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000255006.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000440354.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000459721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000465815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000465815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000467569.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000484638.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
ENST00000488077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_001242581.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000006861.hg.1 |
NM_018993.3 |
Refseq |