This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00609802_m1
See other GCKR GE Assays ›
Gene Symbol:
GCKR
Gene Name:
glucokinase (hexokinase 4) regulator
Gene Aliases:
FGQTL5, GKRP
Chromosome Location:
Chr.2: 27496839 - 27523689 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00609802_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCKR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001486.3 NP_001477.2 3 - 4 357 63
XM_011532763.1 XP_011531065.1 3 - 4 357 63
GenBank mRNA AK291846.1 - 3 - 4 346 63
AK298448.1 - 3 - 4 348 63
AK303784.1 - 3 - 4 348 63
AK309599.1 - 3 - 4 313 63
BC130481.1 - 3 - 4 337 40146512 63
BC130483.1 - 3 - 4 337 40146514 63
DA011063.1 - 3 - 4 357 63
Z48475.1 - 3 - 4 354 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200007070.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02000176.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK303784.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GCKR

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus, translocation
carbohydrate metabolic process
response to fructose
regulation of glucose transport
negative regulation of glucokinase activity
urate metabolic process
triglyceride homeostasis
protein binding
carbohydrate binding
fructose-6-phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02002369.hg.1 ENST00000264717.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02002369.hg.1 ENST00000424318.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02002369.hg.1 NM_001486.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02002369.hg.1 TC02000176.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000411584.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 ENST00000478147.1 Ensembl
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay TC0200007070.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000264717.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000417872.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000453813.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 ENST00000472290.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 NM_001486.3 Refseq
Affymetrix Human Clariom D Assay PSR0200148545.hg.1 TC0200007070.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0