This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00609758_g1
See other CELSR3 GE Assays ›
Gene Symbol:
CELSR3
Gene Name:
cadherin EGF LAG seven-pass G-type receptor 3
Gene Aliases:
ADGRC3, CDHF11, EGFL1, FMI1, HFMI1, MEGF2, RESDA1
Chromosome Location:
Chr.3: 48636463 - 48662915 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00609758_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CELSR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001407.2 NP_001398.2 11 - 12 6030 107
GenBank mRNA AF231023.1 - 11 - 12 6030 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013973.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03003382.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CELSR3

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
regulation of protein phosphorylation
homophilic cell adhesion via plasma membrane adhesion molecules
G-protein coupled receptor signaling pathway
axonal fasciculation
regulation of protein localization
dopaminergic neuron axon guidance
serotonergic neuron axon guidance
cilium assembly
Wnt signaling pathway, planar cell polarity pathway
G-protein coupled receptor activity
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03024338.hg.1 ENST00000164024.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03024338.hg.1 ENST00000544264.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03024338.hg.1 NM_001407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03024338.hg.1 TC03003382.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0300207456.hg.1 ENST00000164024.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300207456.hg.1 NM_001407.2 Refseq
Affymetrix Human Clariom D Assay PSR0300207456.hg.1 ENST00000164024.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300207456.hg.1 NM_001407.2 Refseq
Affymetrix Human Clariom D Assay PSR0300207456.hg.1 TC0300013973.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0