This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00609514_m1
See other CD247 GE Assays ›
Gene Symbol:
CD247
Gene Name:
CD247 molecule
Gene Aliases:
CD3-ZETA, CD3H, CD3Q, CD3Z, IMD25, T3Z, TCRZ
Chromosome Location:
Chr.1: 167430640 - 167518610 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00609514_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD247
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000734.3 NP_000725.1 6 - 7 535 68
NM_198053.2 NP_932170.1 6 - 7 538 68
XM_011510144.2 XP_011508446.1 7 - 8 606 68
XM_011510145.1 XP_011508447.1 7 - 8 612 68
XM_017002800.1 XP_016858289.1 7 - 8 597 68
XM_017002801.1 XP_016858290.1 7 - 8 592 68
GenBank mRNA AF228312.1 - 5 - 6 345 68
AK128376.1 - 5 - 6 1866 68
AK223325.1 - 6 - 7 526 68
AK313946.1 - 6 - 7 464 68
AL557555.3 - 6 - 7 535 68
BC025703.1 - 6 - 7 454 5222574 68
EU117371.1 - 2 - 3 90 68
EU117372.1 - 2 - 3 90 68
EU117373.1 - 2 - 3 90 68
EU118788.1 - 2 - 3 90 68
EU118789.1 - 2 - 3 90 68
EU118790.1 - 2 - 3 90 68
EU118791.1 - 2 - 3 90 68
EU118792.1 - 2 - 3 90 68
EU118793.1 - 2 - 3 90 68
EU597245.1 - 4 - 5 237 68
J04132.1 - 6 - 7 464 68
X55510.1 - 6 - 7 434 68
X55511.1 - 6 - 7 434 68
X55512.1 - 6 - 7 437 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100016302.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003479.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD247

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

viral process
T cell costimulation
Fc-gamma receptor signaling pathway involved in phagocytosis
regulation of defense response to virus by virus
regulation of immune response
T cell receptor signaling pathway
transmembrane signaling receptor activity
protein binding
identical protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01054117.hg.1 ENST00000362089.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01054117.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01054117.hg.1 ENST00000483825.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01054117.hg.1 NM_000734.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01054117.hg.1 NM_198053.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01054117.hg.1 TC01003479.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay PSR0100224008.hg.1 TC0100016302.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000362089.9 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000392122.3 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000470379.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000476733.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000479979.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000483825.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 ENST00000485089.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_000734.3 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq
Affymetrix Human Clariom D Assay TC0100016302.hg.1 NM_198053.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0